Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 30/GJB6 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310836100UL
Description
Connexin 30/GJB6 Polyclonal specifically detects Connexin 30/GJB6 in Mouse samples. It is validated for Western Blot.Specifications
Connexin 30/GJB6 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
connexin 30, connexin-30, Cx30, CX30gap junction protein, beta 6, DFNA3B, DFNB1B, ectodermal dysplasia 2, hidrotic (Clouston syndrome), ED2, EDH, gap junction beta-6 protein, gap junction protein, beta 6 (connexin 30), gap junction protein, beta 6, 30kDa, HEDDFNA3 | |
The immunogen is a synthetic peptide directed towards the C terminal region of mouse Connexin 30/GJB6 (NP_001010937.1). Peptide sequence ISASVICMLLNVAELCYLLLKLCFRRSKRTQAQRNHPNHALKESKQNEMN | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
10804 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction