Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 31/GJB3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Connexin 31/GJB3 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Connexin 31/GJB3 Polyclonal specifically detects Connexin 31/GJB3 in Mouse samples. It is validated for Western Blot.Specifications
Connexin 31/GJB3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
connexin 31, connexin-31, Cx31, CX31MGC102938, DFNA2, DFNA2B, EKV, erythrokeratodermia variabilis, FLJ22486, gap junction beta-3 protein, gap junction protein, beta 3, 31kD (connexin 31), gap junction protein, beta 3, 31kDa, gap junction protein, beta 3, 31kDa (connexin 31) | |
The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_032152). Peptide sequence MDWKKLQDLLSGVNQYSTAFGRIWLSVVFVFRVLVYVVAAERVWGDEQKD | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
2707 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title