Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 32/GJB1 Antibody (GJB1/1753), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB008092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008092 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008092 Supplier Novus Biologicals Supplier No. NBP254359DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Connexin 32/GJB1 Monoclonal specifically detects Connexin 32/GJB1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Connexin 32/GJB1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GJB1/1753
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked)
Gene Symbols GJB1
Host Species Mouse
Immunogen Recombinant human Connexin 32/GJB1 protein (Uniprot: P08034)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This Ab recognizes a protein of 27-32kDa, identified as Connexin 32. The connexin family of proteins forms hexameric complexes called 'connexons' that facilitate movement of low molecular weight proteins between cells via gap junctions. Connexin proteins share a common topology of four transmembrane therefore, one connexin usually cannot fully substitute for another.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.