Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 32/GJB1 Antibody (M12.13), Novus Biologicals™
SDP

Catalog No. NB25338120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25338120 20 μg
NB253381100 100 μg
NB253381200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25338120 Supplier Novus Biologicals Supplier No. NBP25338120UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Connexin 32/GJB1 Monoclonal specifically detects Connexin 32/GJB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Connexin 32/GJB1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), SDS-Page
Classification Monoclonal
Clone M12.13
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked)
Gene Symbols GJB1
Host Species Mouse
Immunogen Rat junctional complexes
Molecular Weight of Antigen 29 kDa
Purification Method Protein G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2705
Target Species Human, Mouse, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.