Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 36/GJD2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159254 Supplier Novus Biologicals Supplier No. NBP159254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Connexin 36/GJD2 Polyclonal specifically detects Connexin 36/GJD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Connexin 36/GJD2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:1000
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UKL4
Gene Alias Connexin-36, Cx36, CX36connexin 36, Gap junction alpha-9 protein, gap junction delta-2 protein, gap junction protein, alpha 9, 36kDa, gap junction protein, delta 2, 36kDa, GJA9connexin-36, MGC138315, MGC138319
Gene Symbols GJD2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GJD2(gap junction protein, delta 2, 36kDa) The peptide sequence was selected from the middle region of GJD2. Peptide sequence ELNHLGWRKIKLAVRGAQAKRKSIYEIRNKDLPRVSVPNFGRTQSSDSAY.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57369
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Chicken: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.