Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 43/GJA1 Antibody, Novus Biologicals™
SDP

Catalog No. NB295269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295269 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295269 Supplier Novus Biologicals Supplier No. NBP324691

Rabbit Polyclonal Antibody

Connexin 43/GJA1 Polyclonal antibody specifically detects Connexin 43/GJA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Connexin 43/GJA1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100
Formulation PBS
Gene Alias connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 107-138 amino acids from the N-terminal region of human Connexin 43/GJA1.
Purification Method Protein G purified
Quantity 400 μL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2697
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.