Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 45/GJC1 Antibody (5B9.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019452 0.025 mg
NBP229754 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019452 Supplier Novus Biologicals Supplier No. NBP2297540.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Connexin 45/GJC1 Monoclonal specifically detects Connexin 45/GJC1 in Human, Rabbit, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Connexin 45/GJC1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5B9.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation 20mM Phosphate (pH 7.6) and 0.25M NaCl with 0.1% Sodium Azide
Gene Accession No. P36383
Gene Alias connexin 45, Connexin-45, CX45, DKFZp686P0738, Gap junction alpha-7 protein, gap junction gamma-1 protein, gap junction protein, alpha 7, 45kDa, gap junction protein, gamma 1, 45kDa, GJA7
Gene Symbols GJC1
Host Species Mouse
Immunogen The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-377 of human connexin 45.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10052
Test Specificity Recognizes Human Connexin 45, also confirmed to work on rat C6 cells.
Target Species Human, Rabbit, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.