Learn More
Description
Specifications
Specifications
| Antigen | Connexin 50/GJA8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | P48165 |
| Gene Alias | alpha 8, 50kD (connexin 50), CAE, Connexin-50, Cx50, CZP1, gap junction alpha-8 protein, gap junction membrane channel protein alpha-8, gap junction protein, alpha 8, 50kDa, gap junction protein, alpha 8, 50kDa (connexin 50), lens intrinsic membrane protein MP70, MP70 |
| Gene Symbols | GJA8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
