Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Connexin 58/GJA9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Connexin 58/GJA9 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Connexin 58/GJA9 Polyclonal specifically detects Connexin 58/GJA9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Connexin 58/GJA9 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
connexin 59, connexin-58, Connexin-59, CX58, Cx59, CX59gap junction alpha 10, Gap junction alpha-10 protein, gap junction alpha-9 protein, gap junction protein, alpha 10, 59kDa, gap junction protein, alpha 9, 59kDa, GJA10, MGC50985 | |
GJA9 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
P57773 | |
81025 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LNGNQLMEKRETEGKDSKRNYYSRGHRSIPGVAIDGENNMRQSPQTVFSLPANCDWKPRWLRATWGSSTEHENRGSPPKGN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title