Learn More
Description
Specifications
Specifications
| Antigen | Contactin-1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | contactin 1, contactin-1, F3, Glycoprotein gp135, GP135, Neural cell surface protein F3 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Contactin-1 (NP_001834.2; Met 1-993). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
