Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contactin-2/TAG1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB320791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320791 Supplier Novus Biologicals Supplier No. NBP298381APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Contactin-2/TAG1 Polyclonal antibody specifically detects Contactin-2/TAG1 in Mouse samples. It is validated for ELISA

Specifications

Antigen Contactin-2/TAG1
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias Axonal glycoprotein TAG-1, Axonin-1, axonin-1 cell adhesion molecule, AXT, contactin 2 (axonal), contactin 2 (transiently expressed), contactin-2, DKFZp781D102, FLJ37193, MGC157722, TAG1, TAG-1, TAX, TAX-1, TAX1FLJ42746, Transient axonal glycoprotein 1, transiently-expressed axonal glycoprotein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Contactin-2/TAG1 (Uniprot#: Q61330; Gln31-Glu1013)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6900
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.