Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contactin-3 Antibody (006), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB308052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308052 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308052 Supplier Novus Biologicals Supplier No. NBP289312MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Contactin-3 Monoclonal antibody specifically detects Contactin-3 in Human samples. It is validated for ELISA

Specifications

Antigen Contactin-3
Applications ELISA
Classification Monoclonal
Clone 006
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias contactin 3 (plasmacytoma associated)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Contactin-3 (Met1-Ser1002,708 Asp/Asn; Accession#: NP_065923.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5067
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.