Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contactin-3 Antibody (524518) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB376064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376064 Supplier Novus Biologicals Supplier No. FAB6644A
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Contactin-3 Monoclonal antibody specifically detects Contactin-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Contactin-3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 524518
Conjugate APC
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias contactin 3 (plasmacytoma associated)
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-3, Glu20-Thr1003, Accession # Q07409
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5067
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.