Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contactin-3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB186695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186695 200 μg
NB186696 50 μg
NB186694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB186695 Supplier Novus Biologicals Supplier No. NBP312813200UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Contactin-3 Polyclonal antibody specifically detects Contactin-3 in Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Contactin-3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Formulation 0.2μm filtered solution in PBS without Preservative
Gene Alias contactin 3 (plasmacytoma associated)
Host Species Rabbit
Immunogen Purified, recombinant Rat Contactin-3 (Q62682; Met1-Gly1001)
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5067
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.