Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Contactin 5 Monoclonal Antibody (4C10E3C12), Invitrogen™

Catalog No. p-7240537
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543508 50 μL
PIMA542992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543508 Supplier Invitrogen™ Supplier No. MA543508
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human CNTN5/Contactin-5

The protein encoded by this gene is a member of the immunoglobulin superfamily, and contactin family, which mediate cell surface interactions during nervous system development. This protein is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 5
Applications ELISA
Classification Monoclonal
Clone 4C10E3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CNTN5
Gene Accession No. O94779
Gene Alias 6720426O10Rik; A830025P08Rik; CNTN5; contactin 5; contactin-5; Gm507; hNB-2; HNB-2s; MGC163491; Nb-2; neural adhesion molecule; neural recognition molecule NB-2
Gene Symbols CNTN5
Host Species Mouse
Immunogen Recombinant Human CNTN5 protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53942
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.