Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COP9 Antibody (OTI3E1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP271382FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271382FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271382FR Supplier Novus Biologicals Supplier No. NBP271382FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COP9 Monoclonal specifically detects COP9 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen COP9
Applications Western Blot
Classification Monoclonal
Clone OTI3E1
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis), COP9 signalosome complex subunit 3, CSN3COP9 complex subunit 3, JAB1-containing signalosome subunit 3, SGN3COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 3, Signalosome subunit 3
Gene Symbols COPS3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 224-423 of human COP9 (NP_003644) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8533.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.