Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPB2 Antibody (M3A5), Novus Biologicals™
SDP

Catalog No. NB600102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600102 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600102 Supplier Novus Biologicals Supplier No. NB600102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

COPB2 Monoclonal specifically detects COPB2 in Human, Rat, Avian, Bovine, Canine, Chicken, Hamster, Monkey, Primate, Rabbit samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen COPB2
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Affinity Purification
Classification Monoclonal
Clone M3A5
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification
Gene Alias beta prime subunit, Beta'-coat protein, Beta'-COP, betaprime-COP, coatomer protein complex, subunit beta 2 (beta prime), coatomer subunit beta', p102
Gene Symbols COPB2
Host Species Mouse
Immunogen Microtubule-associated protein from goose brain.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9276
Test Specificity Recognizes an epitope shared by the beta-COP protein (110kD) found in most tissue culture cells, and a high MW doublet of brain microtubule-associated protein (MAP 2, 270-300 kD).
Target Species Human, Rat, Avian, Bovine, Canine, Chicken, Hamster, Monkey, Primate, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.