Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPG Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB058763 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB058763 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB058763 Supplier Novus Biologicals Supplier No. NBP299186C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COPG Polyclonal specifically detects COPG in Human samples. It is validated for Western Blot.

Specifications

Antigen COPG
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias coat protein gamma-cop, coatomer protein complex, subunit gamma, coatomer protein complex, subunit gamma 1, COPG1coatomer subunit gamma, FLJ21068, Gamma-coat protein, gamma-COP
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human COPG.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22820
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.