Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | COPG2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
COPG2 Polyclonal specifically detects COPG2 in Human samples. It is validated for Western Blot.Specifications
COPG2 | |
Polyclonal | |
Rabbit | |
Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
2-COP, coat protein, nonclathrin, gamma-2-cop, coatomer protein complex, subunit gamma 2, coatomer subunit gamma-2, DKFZp761N09121, FLJ11781, Gamma-2-coat protein, gamma-2-COP | |
COPG2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q9UBF2 | |
26958 | |
Synthetic peptides corresponding to COPG2(coatomer protein complex, subunit gamma 2) The peptide sequence was selected from the N terminal of COPG2. Peptide sequence MIKKFDKKDEESGSGSNPFQHLEKSAVLQEARIFNETPINPRRCLHILTK. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title