Learn More
Copper(II) 2-ethylhexanoate
Description
Copper(II) 2-ethylhexanoate is used as pharmaceutical intermediates. Used as drier/additive for paints, coatings and inks. Used as catalystic special applications in lubricant and grease additives, fuel combustion additives to enhance and stabilize the performance of fuels. Used in the the rubber wire coat compound and the brass-coated steel belt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 252°C (decomposition) |
| Color | Blue-Green |
| Quantity | 50 g |
| Solubility Information | Soluble in water,alcohols,dichloroethane,chlorobenzene,and most organic solvents. |
| Formula Weight | 349.96 |
| Physical Form | Powder or Lumps |
| Chemical Name or Material | Copper(II) 2-ethylhexanoate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 205-731-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.