Learn More
Copper(II) fluoride dihydrate
Description
Copper(II) fluoride can also be used to form fluorobenzene from benzene, hydrochloric acid and oxygen. Using a metal fluoride that can successfully oxidize with a methyl bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >130°C (decomposition) |
| Physical Form | Powder |
| Quantity | 250 g |
| UN Number | UN3288 |
| Merck Index | 14,2638 |
| Solubility Information | Slightly soluble in cold water, hydrolyzes to oxyfluoride in hot water |
| Formula Weight | 137.57 (101.54 Anhydrous) |
| Odor | Odorless |
| Density | 2.934 g/mL |
| Chemical Name or Material | Copper(II) fluoride dihydrate |
Safety and Handling
| Hazard Category | H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H331-H302 Toxic if inhaled. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 232-147-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.