Learn More
Copper(II) trifluoromethanesulfonate, 99%
Description
Copper(II) trifluoromethanesulfonate is a mild lewis acid. It is used as catalyst which promotes dehydration of alcohols and diols to alkenes at ambient temperatures. It is widely used to generate carbenoid species from ?-diazo esters and ketones, via in situ reduction to the Cu(I) species. It is also promotes the reaction between diazo esters and imines to give aziridines. It catalyzes syn-selective aldol condensation of (Z)-silyl enol ethers with aldehydes, Friedel-Crafts alkylation, acylation reactions of aromatics and addition of trimethylsilyl cyanide to carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | (decomposition) | 
| Color | Blue to White | 
| Physical Form | Powder | 
| Quantity | 10 g | 
| UN Number | UN3261 | 
| Beilstein | 4028198 | 
| Solubility Information | Soluble in water. | 
| Formula Weight | 361.68 | 
| Percent Purity | ≤99% | 
| Sensitivity | Hygroscopic | 
| Show More | 
Safety and Handling
| Hazard Category | H314 | 
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage.  | 
                                
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c | 
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. | 
| EINECSNumber | 252-300-8 | 
| TSCA | No | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.