Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPR5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP23088425UL
Description
COPR5 Polyclonal specifically detects COPR5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COPR5 | |
Polyclonal | |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9NQ92 | |
COPRS | |
This antibody was developed against a recombinant protein corresponding to amino acids: EAGFATADHSGQERETEKAMDRLARGTQSIPNDSPARGEGTHSEEEGFAMDEEDSDGELNTWELSEGTNCPPKE | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C17orf79, chromosome 17 open reading frame 79, cooperator of PRMT5, COPR5, FLJ21119, HSA272196, Protein TTP1, TTP1 | |
Rabbit | |
Affinity Purified | |
RUO | |
55352 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction