Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPR5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $646.00
Specifications
Antigen | COPR5 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
COPR5 Polyclonal specifically detects COPR5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COPR5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C17orf79, chromosome 17 open reading frame 79, cooperator of PRMT5, COPR5, FLJ21119, HSA272196, Protein TTP1, TTP1 | |
COPRS | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
Q9NQ92 | |
55352 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EAGFATADHSGQERETEKAMDRLARGTQSIPNDSPARGEGTHSEEEGFAMDEEDSDGELNTWELSEGTNCPPKE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title