Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS6 Antibody (OTI4E7), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271648G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271648G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271648G Supplier Novus Biologicals Supplier No. NBP271648G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COPS6 Monoclonal specifically detects COPS6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COPS6
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4E7
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BMP2B, BMP2B1, BMP4, COP9 constitutive photomorphogenic homolog subunit 6 (Arabidopsis), COP9 signalosome complex subunit 6, CSN6COP9 subunit 6 (MOV34 homolog, 34 kD), H_NH0506M12.12, hVIP, JAB1-containing signalosome subunit 6, MOFC11, MOV34 homolog, MOV34 homolog, 34 kD, MOV34-34KD, SGN6, Signalosome subunit 6, Vpr-interacting protein
Gene Symbols COPS6
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 58-327 of human COPS6 (NP_006824) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10980.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.