Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ COQ7 Antibody Pair [HRP]
SDP

Catalog No. p-7183339 Shop All Novus Biologicals Products
Click to view available options
Quantity:
15 Plates

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human COQ7.

Specifications

Assay Range 39.1 pg/mL
Assay Sensitivity 39.06-2500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human COQ7.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CAT5, CLK1, CLK-1, Coenzyme Q biosynthesis protein 7 homolog, coenzyme Q, 7 (rat, yeast) homolog, coenzyme Q7 homolog, ubiquinone (yeast), COQ7 coenzyme Q, 7 homolog ubiquinone, placental protein KG-20, Timing protein clk-1 homolog, ubiquinone biosynthesis protein COQ7 homolog
Gene ID (Entrez) 10229
Gene Symbol COQ7
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 100 ng of recombinant COQ7.
  • Capture Antibody: 0.4 mg/mL of mouse anti-COQ7 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-COQ7 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target COQ7
    Technique 1. Capture Antibody: Dilute to a working concentration of 1.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.