Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COQ7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106348 Supplier Invitrogen™ Supplier No. PA5106348
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total COQ7.

The protein encoded by this gene is similar to a mitochondrial di-iron containing hydroxylase in Saccharomyces cerevisiae that is involved with ubiquinone biosynthesis. Mutations in the yeast gene lead to slower development and longer life span. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COQ7
Gene Accession No. P97478, Q63619, Q99807
Gene Alias 5-demethoxyubiquinone hydroxylase; 5-demethoxyubiquinone hydroxylase, mitochondrial; CAT5; CLK1; CLK-1; Coenzyme q (ubiquinone) biosynthetic enzyme (DHPB methyltransferase) 7; coenzyme Q biosynthesis protein 7 homolog; coenzyme Q7 homolog, ubiquinone; coenzyme Q7 homolog, ubiquinone (yeast); coenzyme Q7, hydroxylase; COQ10D8; COQ7; COQ7 coenzyme Q, 7 homolog ubiquinone; demethyl-Q 7; DMQ hydroxylase; placental protein KG-20; timing protein clk-1 homolog; Ubiquinone biosynthesis monooxygenase COQ7; ubiquinone biosynthesis protein COQ7 homolog
Gene Symbols COQ7
Host Species Rabbit
Immunogen A synthesized peptide derived from human COQ7(Accession Q99807), corresponding to amino acid residues F34-Q84.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10229, 12850, 25249
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.