Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COQ9 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB326661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326661 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326661 Supplier Novus Biologicals Supplier No. NBP299187APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COQ9 Polyclonal antibody specifically detects COQ9 in Human samples. It is validated for Western Blot

Specifications

Antigen COQ9
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias C16orf49DKFZp434K046, chromosome 16 open reading frame 49, coenzyme Q9 homolog (S. cerevisiae), coenzyme Q9 homolog (yeast), DKFZP434K046, ubiquinone biosynthesis protein COQ9, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human COQ9.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57017
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.