Learn More
Eisco™ Coquina Sedimentary Rock Specimen
SCI_ED
Catalog No. p-7203009
Click to view available options
Quantity:
1/Ea.
12/Pk.
Composed of bone and shell fragments, coquina is nearly 100% fossil matter. Held together by a carbonate cement, this skeletal mass forms in low-energy environments such as lagoons and reefs.
  • Correlates well to Next Generation Science Standards Evidence
  • Great for teaching sedimentary facies, paleontology, and stratigraphy
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.52 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.