Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COR 170, Tocris Bioscience™

Catalog No. 475650
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 47-565-0

Supplier: R&D Systems 4756/50

This item is not returnable. View return policy

Selective CB2 receptor inverse agonist

Selective inverse agonist of CB2 receptors (Ki values are 3.8 and >10,000 nM for CB2 and CB1, respectively).

Chemical Identifiers

CAS 1048039-15-1
Molecular Formula C31H36N2O2
Molecular Weight (g/mol) 468.641
InChI Key MSJISJDTJJYBFT-UHFFFAOYSA-N
Synonym n-adamant-1-yl-4-oxo-1-pentyl-6-phenyl-1,4-dihydroquinoline-3-carboxamide
PubChem CID 25033736
IUPAC Name N-(1-adamantyl)-4-oxo-1-pentyl-6-phenylquinoline-3-carboxamide
SMILES CCCCCN1C=C(C(=O)C2=C1C=CC(=C2)C3=CC=CC=C3)C(=O)NC45CC6CC(C4)CC(C6)C5

Specifications

Quantity 50 mg
Formula Weight 468.63
Percent Purity >99%
Chemical Name or Material COR 170
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.