Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corin Antibody (MM0199-9H21) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019099 0.025 mg
NBP212205 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019099 Supplier Novus Biologicals Supplier No. NBP2122050.025MG

Rat Monoclonal Antibody

Corin Monoclonal specifically detects Corin in Human samples. It is validated for Western Blot.

Specifications

Antigen Corin
Applications Western Blot
Classification Monoclonal
Clone MM0199-9H21
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9Y5Q5
Gene Alias atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Gene Symbols CORIN
Host Species Rat
Immunogen Human Corin
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10699
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.