Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corin Antibody (MM0199-9H21), PE, Novus Biologicals™
SDP

Catalog No. NB344281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344281 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344281 Supplier Novus Biologicals Supplier No. NBP212205PE

Rat Monoclonal Antibody

Corin Monoclonal antibody specifically detects Corin in Human samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen Corin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MM0199-9H21
Conjugate PE
Dilution Western Blot, Immunocytochemistry
Formulation PBS
Gene Alias atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Host Species Rat
Immunogen Human recombinant Corin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10699
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.