Description
- Good quality plated brass cork borers with solid brass handles, except for Set of 3 which includes plastic handles.
- Please note: borers are intended for use only with cork stoppers.
Specifications
Specifications
| Type | Cork Borer Set |
| Product Type | Cork Borer Set |
| Quantity | Set of 3 |
Warning: Cancer and reproductive harm