Learn More
Tocris Bioscience™ CORM 3
GSA_VAGREENER_CHOICE
Catalog No. p-5545539
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy
Water-soluble carbon monoxide-releasing molecule
CORM 3 is a water-soluble carbon monoxide-releasing molecule. Suppresses thrombin-induced nitrite release in BV-2 microglia. Protects isolated rats hearts from reperfusion damage in vitro and is cardioprotective in a rat myocardial infarction model in vivo. Also prolongs survival of transplanted hearts in mice. Exhibits anti-inflammatory and vasorelaxant effects.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 10 mg
Product Type CORM 3
Molecular Weight (g/mol) 294.61
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.