Corning™ Microcentrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Specially designed polypropylene tubes protect sensitive samples and provide added security against leakage and contamination. Corning™ Microcentrifuge Tubes withstand forces up to 20,000 x g.
Manufacturer: Corning™ 430915
 View more versions of this product
Catalog No. 05-538-69C
  Check Availability
Add to Cart
    Specifications
    Max. Force 20000 x g
    Product Type Microcentrifuge Tube
    Capacity (Metric) 2mL
    Color Natural
    Material Polypropylene
    Sterile Yes
    Cap Style Attached Screw Cap with silicone O-ring
    Shape Conical Self-Standing
    Closure Type Screw
    • Screw caps feature one-handed capping and uncapping
    • Plug-seal top
    • Standard conical bottom or free-standing skirted bottom
    • Unattached rim-seal screw cap or attached-loop screw cap with silicone O-ring for optimum cap-to-vessel seal
    • All tubes and caps sterilized
  • Description & Specifications
Related Products
Product Content Feedback