Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Storage Mat Applicator
Shop All Corning Products
Click to view available options
No. per Pack:
1 Ea.
Description
Storage Mat Applicators evenly apply storage mats to Corning 96 well polypropylene plates, blocks, and 96 well polystyrene plates.
Specifications
Specifications
| Color | Black |
| For Use With (Application) | For high-volume sample processing |
| No. per Pack | 1 Ea. |
| Product Type | Storage Mat Applicator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction