Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cornulin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7061824
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3607SP 25 μg
AF3607 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3607SP Supplier R&D Systems Supplier No. AF3607SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Cornulin Polyclonal specifically detects Cornulin in Human samples. It is validated for Western Blot.

Specifications

Antigen Cornulin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UBG3
Gene Alias 53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, C1orf10, chromosome 1 open reading frame 10, cornulin, CRNN, DRC1, Gm1015, PDRC1, PDRC1,53 kDa putative calcium-binding protein, SEP53, SEP53C1orf10, Squamous epithelial heat shock protein 53, Tumor-related protein
Gene Symbols CRNN
Host Species Goat
Immunogen E. coli-derived recombinant human Cornulin Met1-Ser140 Accession # Q9UBG3
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49860
Test Specificity Detects human Cornulin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.