Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CORO7 Rabbit anti-Human, Mouse, Rat, Bovine, Orangutan, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP260707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260707 Supplier Novus Biologicals Supplier No. NBP260707

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

CORO7 Polyclonal specifically detects CORO7 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen CORO7
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 20 ul/mg lysate
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias coronin-7, 0610011B16Rik, coronin 7, CRN7, POD1
Gene Symbols CORO7
Host Species Rabbit
Immunogen between 875 and 925
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 79585
Target Species Human, Mouse, Rat, Bovine, Orangutan
Content And Storage Store at 4degreesC.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.