Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronavirus Antibody (5A4), Novus Biologicals™
SDP

Catalog No. NB11016941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11016941 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11016941 Supplier Novus Biologicals Supplier No. NB11016941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coronavirus Monoclonal specifically detects Coronavirus in Virus samples. It is validated for ELISA.

Specifications

Antigen Coronavirus
Applications ELISA, Hemagglutination Assay
Classification Monoclonal
Clone 5A4
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Gene Alias Coronaviridae
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with bovine corona virus.
Purification Method Protein G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Bovine corona virus surface antigen
Target Species Bovine
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.