Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronavirus Antibody (FIPV3-70), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB10064754W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064754W 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064754W Supplier Novus Biologicals Supplier No. NB10064754AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coronavirus Monoclonal specifically detects Coronavirus in Virus samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Coronavirus
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone FIPV3-70
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Coronaviridae
Host Species Mouse
Immunogen Coronavirus cocktail
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64754 reacts with feline infectious peritonitis virus (FIPV) type 1 and 2 and is known to be specific for the nucleocapsid. It is also known to react with canine coronavirus (CCV), pig coronavirus transmissible gastroenteritis virus (TGEV) and ferret coronavirus. Some specific activity has been detected against bovine coronavirus (BCV). Clone FIPV3-70 exhibits negative reactivity against Feline Leukemia virus, Feline Immunodeficiency virus, Feline Calcivirus, Feline Herpes virus, Canine Adenovirus (type 2), Canine Distemper virus, Canine Parvovirus and Canine Parainfluenza virus.
Reconstitution Protein A purified
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.