Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin-1a Antibody (OTI1A5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP271680V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271680V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271680V Supplier Novus Biologicals Supplier No. NBP271680V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coronin-1a Monoclonal specifically detects Coronin-1a in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Coronin-1a
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A5
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias clipin-A, CLIPINA, CORO1, coronin, actin binding protein, 1A, coronin, actin-binding protein, 1A, coronin-1, coronin-1A, Coronin-like protein A, Coronin-like protein p57, FLJ41407, HCORO1, MGC117380, p57, TACOCLABP, Tryptophan aspartate-containing coat protein
Gene Symbols CORO1A
Host Species Mouse
Immunogen Full length recombinant protein of human Coronin-1a (NP_009005) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11151.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.