Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin-1a Antibody (OTI1A5), Novus Biologicals™
SDP

Catalog No. NBP202779 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202779 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202779 Supplier Novus Biologicals Supplier No. NBP202779
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coronin-1a Monoclonal specifically detects Coronin-1a in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Coronin-1a
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Gene Alias clipin-A, CLIPINA, CORO1, coronin, actin binding protein, 1A, coronin, actin-binding protein, 1A, coronin-1, coronin-1A, Coronin-like protein A, Coronin-like protein p57, FLJ41407, HCORO1, MGC117380, p57, TACOCLABP, Tryptophan aspartate-containing coat protein
Gene Symbols CORO1A
Host Species Mouse
Immunogen Full length human recombinant protein of human CORO1A(NP_009005) produced in HEK293T cell.
Molecular Weight of Antigen 50.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11151
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.