Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin-2B Antibody, Novus Biologicals™
SDP

Catalog No. NBP215969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215969 Supplier Novus Biologicals Supplier No. NBP215969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Coronin-2B Polyclonal antibody specifically detects Coronin-2B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Coronin-2B
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.96mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UQ03
Gene Alias clipin C, Clipin-C, CLIPINC, coronin, actin binding protein, 2B, coronin, actin-binding protein, 2B, coronin, actin-binding, 2B, coronin-2B, Coronin-like protein C, KIAA0925clipin-C, Protein FC96
Gene Symbols CORO2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Coronin 2B. The exact sequence is proprietary.
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10391
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Xenopus laevis (87%), Xenopus tropicalis (86%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.