Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coronin 3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227289SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227289SS 0.025 mg
NBP227289 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227289SS Supplier Novus Biologicals Supplier No. NBP227289SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Coronin 3 Polyclonal specifically detects Coronin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Coronin 3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:50-1:100
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Alias coronin 1C, coronin, actin binding protein, 1C, coronin, actin-binding protein, 1C, coronin-1C, Coronin-3, CRNN4, HCRNN4
Gene Symbols CORO1C
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 407-426 of human CORO1C.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23603
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.