Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Corrugated Cardboard Box

Fisherbrand Corrugated Cardboard Box for shipping, packaging and storage

Specifications

Quantity 20
Color Brown
Height (English) Exterior 4.5 in.
Material 32 ECT Cardboard
View More Specs

Products 4
Catalog Number Dimensions (LxWxH) Price Quantity & Availability  
Catalog Number Dimensions (LxWxH) Price Quantity & Availability  
07-100-114
View Documents
8 x 8 in. N/A N/A N/A
07-100-115
View Documents
10 in. x 10 in. N/A N/A N/A
07-100-116
View Documents
12 x 12 in. N/A N/A N/A
07-100-117
View Documents
14 in. x 14 in. N/A N/A N/A
Description

Description

Ships flat to reduce shipping and storage cost

Order Info

Case quantity only 20 per case

Warranty and Services

Warranty against defects in material or workmanship.

Specifications

Specifications

20
Brown
4.5 in.
32 ECT Cardboard
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.