Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortactin, p Tyr466 Antibody, Novus Biologicals™
SDP

Catalog No. NB10081871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10081871 0.05 mL
NB017830 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10081871 Supplier Novus Biologicals Supplier No. NB10081871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cortactin (p Tyr466) Polyclonal specifically detects Cortactin (p Tyr466) in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Cortactin (p Tyr466)
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. Q14247
Gene Alias 1110020L01Rik, Amplaxin, cortactin, ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 srcsubstrate), EMS1amplaxin, FLJ34459, Oncogene EMS1, src substrate cortactin
Gene Symbols CTTN
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from Human Cortactin around the phosphorylation site of tyrosine 466 (P-V-YP-E-T).
Molecular Weight of Antigen 85 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2017
Test Specificity Detects endogenous levels of Cortactin only when phosphorylated at tyrosine 466.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.