Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cortactin, p Tyr466 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $402.00
Specifications
Antigen | Cortactin (p Tyr466) |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017830
![]() |
Novus Biologicals
NB100818710.025ML |
0.025 mL |
Each for $243.50
|
|
|||||
NB10081871
![]() |
Novus Biologicals
NB10081871 |
0.05 mL |
Each for $402.00
|
|
|||||
Description
Cortactin Polyclonal specifically detects Cortactin in Human, Mouse samples. It is validated for Western Blot.Specifications
Cortactin (p Tyr466) | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Cytoskeleton Markers, Phospho Specific | |
Q14247 | |
2017 | |
IgG | |
Affinity Purified | |
85 kDa |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Human | |
1110020L01Rik, Amplaxin, cortactin, ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 srcsubstrate), EMS1amplaxin, FLJ34459, Oncogene EMS1, src substrate cortactin | |
CTTN | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title