Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corticosterone Antibody, Novus Biologicals™
SDP

Catalog No. NB100624805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100624805 0.5 mL
NB017484 0.1 mL
NB10062480 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB100624805 Supplier Novus Biologicals Supplier No. NB100624800.5ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Corticosterone Polyclonal specifically detects Corticosterone in Fish, Non-species specific, Rat samples. It is validated for ELISA, Radioimmune Assays (RIA).

Specifications

Antigen Corticosterone
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1/5K-1/20K
Formulation PBS with 0.09% sodium azide
Host Species Rabbit
Immunogen Corticosterone 3 -BSA conjugate.
Quantity 0.5 mL
Primary or Secondary Primary
Test Specificity This product is specific for Corticosterone, a steroid hormone which is produced by the adrenal gland. It is the major glucocorticoid in many species, having a role in stress response and immune reactions. This antibody show minimal cross-reactivity with the following related hormones: 11-Deoxycorticosterone 1% Cortisol 0.1% Progesterone 0.02% Testosterone <0.01%
Target Species Fish, Non-species specific, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.