Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corticosterone Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $706.50
Specifications
Antigen | Corticosterone |
---|---|
Concentration | 1.0 mg/mL |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017484
![]() |
Novus Biologicals
NB100624800.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB100624805
![]() |
Novus Biologicals
NB100624800.5ML |
0.5 mL |
Each for $499.50
|
|
|||||
NB10062480
![]() |
Novus Biologicals
NB10062480 |
1 mL |
Each for $706.50
|
|
|||||
Description
Corticosterone Polyclonal specifically detects Corticosterone in All Species samples. It is validated for ELISA, Radioimmunoassay.Specifications
Corticosterone | |
Polyclonal | |
Rabbit | |
Corticosterone 3 -BSA conjugate. | |
Primary |
1.0 mg/mL | |
Unconjugated | |
All species | |
IgG | |
This product is specific for Corticosterone, a steroid hormone which is produced by the adrenal gland. It is the major glucocorticoid in many species, having a role in stress response and immune reactions. This antibody show minimal cross-reactivity with the following related hormones: 11-Deoxycorticosterone 1% Cortisol 0.1% Progesterone 0.02% Testosterone <0.01% |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title