Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corticotropin Releasing Factor Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241273
Description
Corticotropin Releasing Factor Polyclonal specifically detects Corticotropin Releasing Factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Corticotropin Releasing Factor | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CRH | |
Antibody was raised against a 16 amino acid peptide near the amino terminus of human CRH. The immunogen is located within amino acids 30 - 80 of CRH. Amino Acid Squence: FQPPPQSEQPQQPQAR | |
Affinity Purified | |
RUO | |
1392 | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor | |
Rabbit | |
29 kDa | |
0.1 mg | |
Primary | |
CRH antibody is human specific. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction